accessibility menu, dialog, popup

UserName

CAS RN 59-05-2

IUPAC Name: (2S)-2-[(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}phenyl)formamido]pentanedioic acid
Synonyms: methotrexate ledertrexate hdmtx arbitrexate methotrexatum 甲氨蝶呤 abitrexate antifolan MTX folex
Molecular Weight (g/mol): 454.45
Molecular Formula: C20H22N8O5
InChi Key: FBOZXECLQNJBKD-ZDUSSCGKSA-N
SMILES: CN(CC1=CN=C2N=C(N)N=C(N)C2=N1)C1=CC=C(C=C1)C(=O)N[C@@H](CCC(O)=O)C(O)=O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)