accessibility menu, dialog, popup

UserName

CAS RN 57-71-6

IUPAC Name: (3E)-3-(hydroxyimino)butan-2-one
Synonyms: diacetylmonoxime|diacetyl monooxime|DAM|dam (oxime)|isonitrosoethyl methyl ketone|(E)-diacetyl monoxime
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: FSEUPUDHEBLWJY-HWKANZROSA-N
SMILES: CC(=O)C(\C)=N\O
Boiling Point 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)