missing translation for 'onlineSavingsMsg'
Learn More

Diacetyl Monoxime 98.0+%, TCI America™

Catalog No. B068325G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B068325G 25 g
B0683100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B068325G Supplier TCI America Supplier No. B068325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-71-6
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00002116
InChI Key FSEUPUDHEBLWJY-HWKANZROSA-N
Synonym 2,3-butanedione monoxime, diacetyl monoxime, diacetylmonoxime, biacetyl monoxime, biacetyl monooxime, diacetyl monooxime, diacetylmonooxime, 2-oximino-3-butanone, 3-oximino-2-butanone, 2,3-butanedione-2-monoxime
PubChem CID 6409633
ChEBI CHEBI:4480
IUPAC Name (3E)-3-(hydroxyimino)butan-2-one
SMILES CC(=O)C(\C)=N\O

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 186°C
Quantity 25 g
Formula Weight 101.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diacetyl Monoxime
EINECSNumber (9)-0538
RTECSNumber EK3150000
TSCA Yes