missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione monoxime, 99%

Catalog No. AAA1433936
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1433936 500 g
AAA1433922 100 g
AAA143390E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1433936 Supplier Thermo Scientific Chemicals Supplier No. A1433936

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol

2,3-Butanedione monoxime is a reversible inhibitor of myosin ATPase. It is also used as a reagent for colorimetric determination of urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Butanedione monoxime is a reversible inhibitor of myosin ATPase. It is also used as a reagent for colorimetric determination of urea.

Solubility
Soluble in water (50g/L at 20°C)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 57-71-6
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00002116
InChI Key FSEUPUDHEBLWJY-HWKANZROSA-N
Synonym 2,3-butanedione monoxime, diacetyl monoxime, diacetylmonoxime, biacetyl monoxime, biacetyl monooxime, diacetyl monooxime, diacetylmonooxime, 2-oximino-3-butanone, 3-oximino-2-butanone, 2,3-butanedione-2-monoxime
PubChem CID 6409633
ChEBI CHEBI:4480
IUPAC Name (3E)-3-hydroxyiminobutan-2-one
SMILES CC(=O)C(\C)=N\O

Specifications

Melting Point 74°C to 77°C
Boiling Point 185°C to 186°C
Quantity 500 g
Beilstein 605582
Sensitivity Hygroscopic
Solubility Information Soluble in water (50g/L at 20°C)
Formula Weight 101.11
Percent Purity 99%
Chemical Name or Material 2,3-Butanedione monoxime
EINECSNumber 200-348-5
RTECSNumber EK3150000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only