accessibility menu, dialog, popup

UserName

O(S)ON(R)(S)(R)(R)(S)N

CAS RN 57-24-9

IUPAC Name: (1R,11S,18S,20R,21R,22S)-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-9-one
Synonyms: strychnine caswell no. 805 certox strychnine (substance) dolco mouse cereal strychnine and its salts rcra waste number P108 strychnidin-10-one hare-rid strychnine product (product)
Molecular Weight (g/mol): 334.42
Molecular Formula: C21H22N2O2
InChi Key: QMGVPVSNSZLJIA-FVWCLLPLSA-N
SMILES: O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61
Percent Purity 
  • (1)
  • (1)
  • (6)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Physical Form 
  • (1)
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Grade 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (4)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)