missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Strychnine, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Strychnine |
| CAS | 57-24-9 |
| Product Line | phyproof |
| Linear Formula | C21H22N2O2 |
| MDL Number | MFCD00005941 |
| Certifications/Compliance | ISO 17034 |
| Melting Point | 284°C to 286°C |
| Synonym | (-)-Strychnine |
| Type | Reference Substance |
| Molecular Weight (g/mol) | 334.41 |
| Show More |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG