missing translation for 'onlineSavingsMsg'
Learn More

Strychnine, Free Base, 98%, Spectrum™ Chemical

Catalog No. 18614124
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614124 5 g Amber Glass Bottle
18614123 25 g Amber Glass Bottle
18614122 100 g Amber Glass Bottle
18614121 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614124 Supplier Spectrum Chemical Mfg Cor Supplier No. ST1425GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-24-9

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 5 g
Molecular Weight (g/mol) 334.4
Percent Purity 98%
Grade Not Available
Packaging Amber Glass Bottle
Chemical Name or Material Strychnine, Free Base

CAUTION: For manufacturing or laboratory use only. Not for food, drug or pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.