accessibility menu, dialog, popup

UserName

CAS RN 55011-44-4

CH3SOOH2NH3CNH2

CAS RN 55011-44-4

IUPAC Name: 5,11-diamino-4,12-dimethyl-8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene-8,8-dione
Synonyms: 5,11-diamino-4,12-dimethyl-8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene-8,8-dione
Molecular Weight (g/mol): 274.34
Molecular Formula: C14H14N2O2S
InChi Key: OJSPYCPPVCMEBS-UHFFFAOYSA-N
SMILES: CC1=C(N)C=C2C(=C1)C1=CC(C)=C(N)C=C1S2(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results