accessibility menu, dialog, popup

UserName

H3COCH3

CAS RN 542-50-7

IUPAC Name: heptacosan-14-one
Synonyms: 14-heptacosanone
Molecular Weight (g/mol): 394.73
Molecular Formula: C27H54O
InChi Key: VCZMOZVQLARCOE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)CCCCCCCCCCCCC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)