missing translation for 'onlineSavingsMsg'
Learn More

14-Heptacosanone, 97%

Catalog No. AAB2019114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2019114 25 g
AAB2019106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2019114 Supplier Thermo Scientific Chemicals Supplier No. B2019114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 542-50-7 | C27H54O | 394.728 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 542-50-7
Molecular Formula C27H54O
Molecular Weight (g/mol) 394.728
MDL Number MFCD00026598
InChI Key VCZMOZVQLARCOE-UHFFFAOYSA-N
Synonym 14-heptacosanone, ditridecyl ketone, myristone, unii-fq5vc928nn, fq5vc928nn, acmc-209leu, 14-heptacosanon, di-n-tridecyl ketone
PubChem CID 10955
IUPAC Name heptacosan-14-one
SMILES CCCCCCCCCCCCCC(=O)CCCCCCCCCCCCC

Specifications

Melting Point 76°C to 79°C
Quantity 25 g
Beilstein 1796794
Formula Weight 394.73
Percent Purity 97%
Chemical Name or Material 14-Heptacosanone
EINECSNumber 208-815-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only