missing translation for 'onlineSavingsMsg'
Learn More

14-Heptacosanone 95.0+%, TCI America™

Catalog No. H051225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H051225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H051225G Supplier TCI America Supplier No. H051225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 542-50-7
Molecular Formula C27H54O
Molecular Weight (g/mol) 394.728
MDL Number MFCD00026598
InChI Key VCZMOZVQLARCOE-UHFFFAOYSA-N
Synonym 14-heptacosanone, ditridecyl ketone, myristone, unii-fq5vc928nn, fq5vc928nn, acmc-209leu, 14-heptacosanon, di-n-tridecyl ketone
PubChem CID 10955
IUPAC Name heptacosan-14-one
SMILES CCCCCCCCCCCCCC(=O)CCCCCCCCCCCCC

Specifications

Melting Point 78°C
Quantity 25 g
Formula Weight 394.73
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 14-Heptacosanone
EINECSNumber (2)-2475
TSCA No