CAS RN 54-21-7
Sodium Salicylate (Powder/Certified), Fisher Chemical
CAS: 54-21-7 Molecular Formula: C7H5NaO3 Molecular Weight (g/mol): 160.104 MDL Number: MFCD00002440 InChI Key: ABBQHOQBGMUPJH-UHFFFAOYSA-M Synonym: sodium salicylate,sodium 2-hydroxybenzoate,salsonin,clin,enterosalicyl,enterosalil,entrosalyl,glutosalyl,kerasalicyl,magsalyl PubChem CID: 16760658 ChEBI: CHEBI:9180 IUPAC Name: sodium;2-hydroxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
Sodium Salicylate (Small Crystals, Free Flowing/USP), Fisher Chemical™
CAS: 54-21-7 Molecular Formula: C7H5NaO3 Molecular Weight (g/mol): 160.104 MDL Number: MFCD00002440 InChI Key: ABBQHOQBGMUPJH-UHFFFAOYSA-M Synonym: sodium salicylate,sodium 2-hydroxybenzoate,salsonin,clin,enterosalicyl,enterosalil,entrosalyl,glutosalyl,kerasalicyl,magsalyl PubChem CID: 16760658 ChEBI: CHEBI:9180 IUPAC Name: sodium;2-hydroxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
Sodium salicylate, 99%
CAS: 54-21-7 Molecular Formula: C7H5NaO3 Molecular Weight (g/mol): 160.104 MDL Number: MFCD00002440 InChI Key: ABBQHOQBGMUPJH-UHFFFAOYSA-M Synonym: sodium salicylate,sodium 2-hydroxybenzoate,salsonin,clin,enterosalicyl,enterosalil,entrosalyl,glutosalyl,kerasalicyl,magsalyl PubChem CID: 16760658 ChEBI: CHEBI:9180 IUPAC Name: sodium;2-hydroxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
Salicylic acid, sodium salt, 99+%, Thermo Scientific Chemicals
CAS: 54-21-7 Molecular Formula: C7H5NaO3 Molecular Weight (g/mol): 160.104 MDL Number: MFCD00002440 InChI Key: ABBQHOQBGMUPJH-UHFFFAOYSA-M Synonym: sodium salicylate,sodium 2-hydroxybenzoate,salsonin,clin,enterosalicyl,enterosalil,entrosalyl,glutosalyl,kerasalicyl,magsalyl PubChem CID: 16760658 ChEBI: CHEBI:9180 IUPAC Name: sodium;2-hydroxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
Sodium Salicylate, Reagent Grade, Reagents
CAS: 54-21-7 Molecular Formula: C7H5NaO3 Molecular Weight (g/mol): 160.10 InChI Key: ABBQHOQBGMUPJH-UHFFFAOYSA-M Synonym: Sodium 2-hydroxybenzoate IUPAC Name: sodium 2-hydroxybenzoate SMILES: [Na+].OC1=CC=CC=C1C([O-])=O
Sodium Salicylate, Crystal, USP, 98-102%, Spectrum™ Chemical
CAS: 54-21-7 Molecular Formula: C7H5NaO3 Molecular Weight (g/mol): 160.10 InChI Key: ABBQHOQBGMUPJH-UHFFFAOYSA-M IUPAC Name: sodium 2-hydroxybenzoate SMILES: [Na+].OC1=CC=CC=C1C([O-])=O
Sodium Salicylate, Crystal, USP, 98-102%, Spectrum™ Chemical
CAS: 54-21-7 Molecular Formula: C7H5NaO3 Molecular Weight (g/mol): 160.10 InChI Key: ABBQHOQBGMUPJH-UHFFFAOYSA-M IUPAC Name: sodium 2-hydroxybenzoate SMILES: [Na+].OC1=CC=CC=C1C([O-])=O
Sodium Salicylate, For Analysis, EMSURE™, MilliporeSigma™
CAS: 54-21-7 Molecular Formula: C7H5NaO3 Molecular Weight (g/mol): 160.104 InChI Key: ABBQHOQBGMUPJH-UHFFFAOYSA-M Synonym: sodium salicylate,sodium 2-hydroxybenzoate,salsonin,clin,enterosalicyl,enterosalil,entrosalyl,glutosalyl,kerasalicyl,magsalyl PubChem CID: 16760658 ChEBI: CHEBI:9180 IUPAC Name: sodium;2-hydroxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]