missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, Crystal, 99%, Spectrum™ Chemical

Catalog No. 18613845
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613845 50 kg Amber Glass Bottle
18613841 125 g Amber Glass Bottle
18613842 500 g Amber Glass Bottle
18613843 2.5 kg Amber Glass Bottle
18613844 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613845 Supplier Spectrum Chemical Mfg Cor Supplier No. S142550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54-21-7

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 50 kg
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Salicylate, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.