missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate (Powder/Certified), Fisher Chemical

Catalog No. S395500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S395500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S395500 Supplier Thermo Fisher Scientific Supplier No. S395 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.104
MDL Number MFCD00002440
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
Synonym sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
PubChem CID 16760658
ChEBI CHEBI:9180
IUPAC Name sodium;2-hydroxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]

Specifications

Melting Point 200°C
Color White
Assay Percent Range ≥99.5 %
Packaging Glass Bottle
Linear Formula 2-HOC6H4COONa
Quantity 500 g
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 160.1
Percent Purity ≥99.5%
Grade Certified
Physical Form Solid
Chemical Name or Material Sodium Salicylate
Show More Show Less