missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Salicylate, For Analysis, EMSURE™, MilliporeSigma™
Description
Recommended Storage
+5°C to +30°C.
Specifications
Specifications
| Melting Point | 250°C (decomposition) |
| Color | White |
| pH | 8.7 to 9.3 (10g/L, H2O, 20°C) |
| Physical Form | Crystals |
| Quantity | 1 kg |
| Solubility Information | 77g/L |
| Formula Weight | 141.96 |
| Percent Purity | >99.0% |
| Grade | Not Available |
| Packaging | Plastic Bottle |
| Show More |
Safety and Handling
| EINECSNumber | 200-198-0 |
| RTECSNumber | VO5075000 |
| Recommended Storage | +5°C to +30°C. |