missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, Crystal, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18606775
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606775 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606775 Supplier Spectrum Chemical Mfg Cor Supplier No. SO205500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO205, 54-21-7, C7H5O3Na

Spectrum™ Chemical Sodium Salicylate, Crystal, USP is a non-steroidal anti-inflammatory drug used in medicine as an antipyretic and analgesic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.10
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
IUPAC Name sodium 2-hydroxybenzoate
SMILES [Na+].OC1=CC=CC=C1C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps