accessibility menu, dialog, popup

UserName

CAS RN 53885-35-1

IUPAC Name: hydrogen 5-[(2-chlorophenyl)methyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine chloride
Synonyms: ticlid ticlopid tiklyd ticlodone
Molecular Weight (g/mol): 300.24
Molecular Formula: C14H15Cl2NS
InChi Key: MTKNGOHFNXIVOS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CC=CC=C1CN1CCC2=C(C1)C=CS2
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results