missing translation for 'onlineSavingsMsg'
Learn More

Ticlopidine hydrochloride, ≥99%, MilliporeSigma™ Supelco™

Catalog No. 111014526
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014526 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014526 Supplier Merck Emd Millipore Supplier No. T66541G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 53885-35-1
Physical Form Neat
Linear Formula C14H14 ClNS · HCl
MDL Number MFCD00079606
Synonym 5-(o-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
Type Analytical Standard
Industry Type Drug Analysis
Quantity 1 g
Formula Weight 300.25
Percent Purity ≥99%
Grade Analytical Standard
Show More Show Less