missing translation for 'onlineSavingsMsg'
Learn More

Ticlopidine Hydrochloride 98.0+%, TCI America™

Catalog No. T31105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31105G 5 g
T311025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31105G Supplier TCI America Supplier No. T31105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53885-35-1
Molecular Formula C14H15Cl2NS
Molecular Weight (g/mol) 300.24
MDL Number MFCD00079606
InChI Key MTKNGOHFNXIVOS-UHFFFAOYSA-N
Synonym ticlopidine hydrochloride, ticlopidine hcl, ticlodix, ticlodone, ticlid, panaldine, tiklid, 5-2-chlorobenzyl-4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, unii-a1l4914fmf
PubChem CID 65335
ChEBI CHEBI:9589
IUPAC Name hydrogen 5-[(2-chlorophenyl)methyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine chloride
SMILES [H+].[Cl-].ClC1=CC=CC=C1CN1CCC2=C(C1)C=CS2

Specifications

Melting Point 210°C
Color White
Quantity 5 g
Formula Weight 300.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ticlopidine Hydrochloride
RTECSNumber XJ9089100
TSCA No