accessibility menu, dialog, popup

UserName

CH3SOOONa

CAS RN 5324-47-0

IUPAC Name: sodium ethanesulfonate
Synonyms: sodium esylate
Molecular Weight (g/mol): 132.11
Molecular Formula: C2H5NaO3S
InChi Key: KQFAFFYKLIBKDE-UHFFFAOYSA-M
SMILES: [Na+].CCS([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)