missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonic acid sodium salt, 98%, may cont. ca 2% water

Catalog No. AAB2182414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2182414 25 g
AAB2182406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2182414 Supplier Thermo Scientific Chemicals Supplier No. B2182414

CAS: 5324-47-0 | C2H5NaO3S | 132.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5324-47-0
Molecular Formula C2H5NaO3S
Molecular Weight (g/mol) 132.109
MDL Number MFCD00066498
InChI Key KQFAFFYKLIBKDE-UHFFFAOYSA-M
Synonym sodium ethanesulfonate, sodium ethanesulphonate, ethanesulfonic acid sodium salt, ethanesulfonic acid, sodium salt, ethane sulfonic acid sodium salt, sodiumethanesulfonate, sodium ethylsulfonate, sodium esylate, sodium ethane sulfonate, ethylsulfonic acid sodium salt
PubChem CID 4192170
IUPAC Name sodium;ethanesulfonate
SMILES CCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point ∼270°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 132.11
Percent Purity 98%
Assay may cont. ≈2% water
Chemical Name or Material Ethanesulfonic acid sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-194-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only