missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethanesulfonate 98.0+%, TCI America™

Catalog No. E02565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02565G 5 g
E025625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E02565G Supplier TCI America Supplier No. E02565G

Chemical Identifiers

CAS 5324-47-0
Molecular Formula C2H5NaO3S
Molecular Weight (g/mol) 132.109
MDL Number MFCD00066498
InChI Key KQFAFFYKLIBKDE-UHFFFAOYSA-M
Synonym sodium ethanesulfonate, sodium ethanesulphonate, ethanesulfonic acid sodium salt, ethanesulfonic acid, sodium salt, ethane sulfonic acid sodium salt, sodiumethanesulfonate, sodium ethylsulfonate, sodium esylate, sodium ethane sulfonate, ethylsulfonic acid sodium salt
PubChem CID 4192170
IUPAC Name sodium;ethanesulfonate
SMILES CCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 270°C
Color White
Quantity 5 g
Formula Weight 132.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Ethanesulfonate