CAS RN 5162-82-3
CAS RN 5162-82-3
IUPAC Name:
3-chloro-4-methylbenzoate
Synonyms:
3-chloro-4-methylbenzoate
Molecular Weight (g/mol):
169.58
Molecular Formula:
C8H6ClO2
InChi Key:
SDKUOEOJAXGCLU-UHFFFAOYSA-M
SMILES:
CC1=CC=C(C=C1Cl)C([O-])=O
1
–
1
of
1
results
1
3-Chloro-4-methylbenzoic Acid 98.0+%, TCI America™
CAS: 5162-82-3 Molecular Formula: C8H7ClO2 Molecular Weight (g/mol): 170.592 MDL Number: MFCD00045841 InChI Key: SDKUOEOJAXGCLU-UHFFFAOYSA-N Synonym: 3-chloro-p-toluic acid,p-toluic acid, 3-chloro,benzoic acid, 3-chloro-4-methyl,3-chloro-4-methyl-benzoic acid,3-chloro-4-methyl benzoic acid,pubchem4569,acmc-1ayqf,4-09-00-01743 beilstein handbook reference,ksc272s4h,3-chloro,4-methylbenzoic acid PubChem CID: 78840 IUPAC Name: 3-chloro-4-methylbenzoic acid SMILES: CC1=C(C=C(C=C1)C(=O)O)Cl