missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. C24625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24625G 5 g
C246225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24625G Supplier TCI America Supplier No. C24625G
Only null left

Chemical Identifiers

CAS 5162-82-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00045841
InChI Key SDKUOEOJAXGCLU-UHFFFAOYSA-N
Synonym 3-chloro-p-toluic acid, p-toluic acid, 3-chloro, benzoic acid, 3-chloro-4-methyl, 3-chloro-4-methyl-benzoic acid, 3-chloro-4-methyl benzoic acid, pubchem4569, acmc-1ayqf, 4-09-00-01743 beilstein handbook reference, ksc272s4h, 3-chloro,4-methylbenzoic acid
PubChem CID 78840
IUPAC Name 3-chloro-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)Cl

Specifications

Melting Point 202°C
Quantity 5 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-methylbenzoic Acid
RTECSNumber XU1650000
TSCA No