accessibility menu, dialog, popup

UserName

H3CBr

CAS RN 51554-94-0

IUPAC Name: 1-bromo-4-nonylbenzene
Synonyms: 1-bromo-4-nonylbenzene
Molecular Weight (g/mol): 283.25
Molecular Formula: C15H23Br
InChi Key: LVCWOFRWEVUMNY-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)