missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-nonylbenzene, 98%

Catalog No. AAH2704209
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2704209 10 g
AAH2704203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2704209 Supplier Thermo Scientific Chemicals Supplier No. H2704209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51554-94-0 | C15H23Br | 283.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51554-94-0
Molecular Formula C15H23Br
Molecular Weight (g/mol) 283.25
MDL Number MFCD00061116
InChI Key LVCWOFRWEVUMNY-UHFFFAOYSA-N
Synonym 1-bromo-4-n-nonylbenzene, bromononylbenzene, 4-nonylbromobenzene, 4-n-nonylbromobenzene, 4-nonylphenyl bromide, acmc-209kuh, 4-bromo-1-nonylbenzene, 1-bromo-4-nonylbenzene
PubChem CID 12603095
IUPAC Name 1-bromo-4-nonylbenzene
SMILES CCCCCCCCCC1=CC=C(Br)C=C1

Specifications

Density 1.115
Boiling Point 107°C (0.2 mmHg)
Flash Point >120°C (248°F)
Refractive Index 1.509
Quantity 10 g
Formula Weight 283.25
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-n-nonylbenzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only