missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-nonylbenzene 95.0+%, TCI America™

Catalog No. B15965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15965G 5 g
B159625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15965G Supplier TCI America Supplier No. B15965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51554-94-0
Molecular Formula C15H23Br
Molecular Weight (g/mol) 283.25
MDL Number MFCD00061116
InChI Key LVCWOFRWEVUMNY-UHFFFAOYSA-N
Synonym 1-bromo-4-n-nonylbenzene, bromononylbenzene, 4-nonylbromobenzene, 4-n-nonylbromobenzene, 4-nonylphenyl bromide, acmc-209kuh, 4-bromo-1-nonylbenzene, 1-bromo-4-nonylbenzene
PubChem CID 12603095
IUPAC Name 1-bromo-4-nonylbenzene
SMILES CCCCCCCCCC1=CC=C(Br)C=C1

Specifications

Color Colorless
Boiling Point 190°C
Quantity 5 g
Formula Weight 283.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-nonylbenzene