accessibility menu, dialog, popup

UserName

CAS RN 5142-23-4

IUPAC Name: 3-methyl-3H-purin-6-amine
Synonyms: 3-methyladenine|3-methyl-adenine|adenine, 3-methyl-|3-methyl-3H-adenine
Molecular Weight (g/mol): 149.16
Molecular Formula: C6H7N5
InChi Key: FSASIHFSFGAIJM-UHFFFAOYSA-N
SMILES: CN1C=NC(N)=C2N=CN=C12
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Product Type 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results