accessibility menu, dialog, popup

UserName

CH3OONCH3H3CH3CO(E)

CAS RN 51145-57-4

IUPAC Name: ethyl (2E)-2-[(dimethylamino)methylidene]-3-oxobutanoate
Synonyms: ethyl (2E)-2-[(dimethylamino)methylidene]-3-oxobutanoate
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: LQSOVGAUOHMPLK-SOFGYWHQSA-N
SMILES: CCOC(=O)C(=C\N(C)C)\C(C)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results