missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Acetyl-3-(dimethylamino)acrylate 95.0+%, TCI America™

Catalog No. E11795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11795G 5 g
E117925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11795G Supplier TCI America Supplier No. E11795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51145-57-4
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD00087563
InChI Key LQSOVGAUOHMPLK-SOFGYWHQSA-N
Synonym ethyl 2-acetyl-3-dimethylamino acrylate, e-ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl n,n-dimethylaminomethylene acetoacetate, ethyl 2-dimethylamino methylidene-3-oxobutanoate, ethyl 2e-2-dimethylamino methylidene-3-oxobutanoate, 2-acetyl-3-dimethylamino-acrylic acid ethyl ester, ethyl 2e-2-acetyl-3-dimethylamino acrylate, ethyl 2e-2-dimethylaminomethylidene-3-oxobutanoate, ethyl2-acetyl-3-dimethylamino acrylate
PubChem CID 2763877
IUPAC Name ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate
SMILES CCOC(=O)C(=CN(C)C)C(=O)C

Specifications

Boiling Point 150°C
Quantity 5 g
Formula Weight 185.22
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Acetyl-3-(dimethylamino)acrylate