missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-acetyl-3-(dimethylamino)acrylate, 95%, Thermo Scientific Chemicals

Catalog No. AAH6407314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6407314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6407314 Supplier Thermo Scientific Chemicals Supplier No. H6407314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51145-57-4 | Molecular Formula: C9H15NO3 | Molecular Weight: 185.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51145-57-4
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD00087563
InChI Key LQSOVGAUOHMPLK-SOFGYWHQSA-N
PubChem CID 2763877
IUPAC Name ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate
SMILES CCOC(=O)C(=CN(C)C)C(=O)C

Specifications

Color Red
Quantity 25 g
Synonym ethyl 2-acetyl-3-dimethylamino acrylate, e-ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl n,n-dimethylaminomethylene acetoacetate, ethyl 2-dimethylamino methylidene-3-oxobutanoate, ethyl 2e-2-dimethylamino methylidene-3-oxobutanoate, 2-acetyl-3-dimethylamino-acrylic acid ethyl ester, ethyl 2e-2-acetyl-3-dimethylamino acrylate, ethyl 2e-2-dimethylaminomethylidene-3-oxobutanoate, ethyl2-acetyl-3-dimethylamino acrylate
Formula Weight 185.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 2-acetyl-3-(dimethylamino)acrylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only