accessibility menu, dialog, popup

UserName

CAS RN 51-28-5

IUPAC Name: 2,4-dinitrophenol
Synonyms: 2,4-dinitrophenol
Molecular Weight (g/mol): 184.11
Molecular Formula: C6H4N2O5
InChi Key: UFBJCMHMOXMLKC-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results