missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 2,4-Dinitrophenol, MilliporeSigma™ Supelco™

Catalog No. 111020162
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020162 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020162 Supplier Merck Emd Millipore Supplier No. 34334250MG
Only null left

2,4-Dinitrophenol is called as an uncoupler of oxidative phosphorylation, which can prevent adenosine triphosphate(ATP) synthesis, but it permits electron transfer to proceed.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 51-28-5
Vapor Density 6.35 (vs. air)
Physical Form Neat
Linear Formula (O2 N)2C6H3 OH
MDL Number MFCD00007115
Melting Point 108°C to 112°C
Synonym alpha-Dinitrophenol; 2,4-DNP; DNP
Type Analytical Standard
Quantity 250 mg
Formula Weight 184.11
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH