accessibility menu, dialog, popup

UserName

CAS RN 50-78-2

IUPAC Name: 2-(acetyloxy)benzoic acid
Synonyms: aspirin A.S.A. empirin solfrin acetosalin ecolen acidum acetylsalicylicum saletin aspirin (substance) easprin dolean ph 8
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: BSYNRYMUTXBXSQ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1C(O)=O
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)