missing translation for 'onlineSavingsMsg'
Learn More

Sigma Pseudo™ Narcotic Scent (Heroin Formulation Powder), MilliporeSigma™ Supelco™

Catalog No. 111001677
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001677 100 g
111001676 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111001677 Supplier Merck Emd Millipore Supplier No. P2548100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 50-78-2
Product Line Sigma Pseudo
Physical Form Powder
MDL Number MFCD00165842
Type Standard
Quantity 100 g
Grade ACS

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC