accessibility menu, dialog, popup

UserName

CAS RN 498-21-5

IUPAC Name: 2-methylbutanedioic acid
Synonyms: methylsuccinic acid
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: WXUAQHNMJWJLTG-UHFFFAOYNA-N
SMILES: CC(CC(O)=O)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)