missing translation for 'onlineSavingsMsg'
Learn More

Methylsuccinic acid, 99%

Catalog No. AAH6096722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6096722 100 g
AAH6096714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6096722 Supplier Thermo Scientific Chemicals Supplier No. H6096722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 498-21-5 | C5H8O4 | 132.115 g/mol

Methylsuccinic acid is used as a flux material and an important dyestuff intermediate. It is also used as an intermediate in medicine and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylsuccinic acid is used as a flux material and an important dyestuff intermediate. It is also used as an intermediate in medicine and pharmaceuticals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature. Moisture sensitive.

Chemical Identifiers

CAS 498-21-5
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00002659
InChI Key WXUAQHNMJWJLTG-UHFFFAOYSA-N
Synonym 2-methylsuccinic acid, methylsuccinic acid, pyrotartaric acid, butanedioic acid, methyl, methyl succinic acid, 1,2-propanedicarboxylic acid, succinic acid, methyl, 2-methyl-butanedioic acid, butanedioic acid, 2-methyl, ccris 6068
PubChem CID 10349
IUPAC Name 2-methylbutanedioic acid
SMILES CC(CC(=O)O)C(=O)O

Specifications

Melting Point 110°C to 115°C
Linear Formula HO2CCH2CH(CH3)CO2H
Quantity 100 g
Beilstein 1722946
Solubility Information Soluble in water.
Formula Weight 132.11
Percent Purity 99%
Chemical Name or Material Methylsuccinic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 207-857-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only