missing translation for 'onlineSavingsMsg'
Learn More

Methylsuccinic Acid 99.0+%, TCI America™

Catalog No. M043025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M043025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M043025G Supplier TCI America Supplier No. M043025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 498-21-5
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00002659
InChI Key WXUAQHNMJWJLTG-UHFFFAOYSA-N
Synonym 2-methylsuccinic acid, methylsuccinic acid, pyrotartaric acid, butanedioic acid, methyl, methyl succinic acid, 1,2-propanedicarboxylic acid, succinic acid, methyl, 2-methyl-butanedioic acid, butanedioic acid, 2-methyl, ccris 6068
PubChem CID 10349
IUPAC Name 2-methylbutanedioic acid
SMILES CC(CC(=O)O)C(=O)O

Specifications

Melting Point 116°C
Color White
Quantity 25 g
Formula Weight 132.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methylsuccinic Acid
EINECSNumber (2)-0903
TSCA Yes