accessibility menu, dialog, popup

UserName

H3COCH3OCH3

CAS RN 494-99-5

IUPAC Name: 1,2-dimethoxy-4-methylbenzene
Synonyms: 1,2-dimethoxy-4-methylbenzene
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: GYPMBQZAVBFUIZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=C1OC
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)