missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxytoluene 99.0+%, TCI America™

Catalog No. D1986500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1986500G 500 g
D198625G 25 g
D1986100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1986500G Supplier TCI America Supplier No. D1986500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 494-99-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00016651
InChI Key GYPMBQZAVBFUIZ-UHFFFAOYSA-N
Synonym 3,4-dimethoxytoluene, homoveratrole, 4-methylveratrole, benzene, 1,2-dimethoxy-4-methyl, 4-methylveratrol, 4-methyl-1,2-dimethoxybenzene, 1,2-dimethoxy-4-methyl-benzene, unii-349x0g2ssf, homoveratrol
PubChem CID 68126
IUPAC Name 1,2-dimethoxy-4-methylbenzene
SMILES CC1=CC(=C(C=C1)OC)OC

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 152.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,4-Dimethoxytoluene
EINECSNumber (3)-0610
TSCA Yes