missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxytoluene, 98%

Catalog No. AAA1555814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1555814 25 g
AAA1555806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1555814 Supplier Thermo Scientific Chemicals Supplier No. A1555814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 494-99-5 | C9H12O2 | 152.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 494-99-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00016651
InChI Key GYPMBQZAVBFUIZ-UHFFFAOYSA-N
Synonym 3,4-dimethoxytoluene, homoveratrole, 4-methylveratrole, benzene, 1,2-dimethoxy-4-methyl, 4-methylveratrol, 4-methyl-1,2-dimethoxybenzene, 1,2-dimethoxy-4-methyl-benzene, unii-349x0g2ssf, homoveratrol
PubChem CID 68126
IUPAC Name 1,2-dimethoxy-4-methylbenzene
SMILES CC1=CC(=C(C=C1)OC)OC

Specifications

Melting Point 25°C to 28°C
Density 1.059
Boiling Point 220°C
Flash Point 84°C (183°F)
Odor Aromatic
Refractive Index 1.528
Quantity 25 g
Beilstein 2045328
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxytoluene
Show More Show Less
EINECSNumber 207-796-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only