accessibility menu, dialog, popup

UserName

CAS RN 4916-55-6

IUPAC Name: hydrogen 3-(bromomethyl)pyridine bromide
Synonyms: hydrogen 3-(bromomethyl)pyridine bromide
Molecular Weight (g/mol): 252.94
Molecular Formula: C6H7Br2N
InChi Key: FNHPUOJKUXFUKN-UHFFFAOYSA-N
SMILES: [H+].[Br-].BrCC1=CC=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)