missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)pyridine Hydrobromide 98.0+%, TCI America™

Catalog No. B21705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21705G 5 g
B217025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21705G Supplier TCI America Supplier No. B21705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4916-55-6
Molecular Formula C6H7Br2N
Molecular Weight (g/mol) 252.937
MDL Number MFCD01321314
InChI Key FNHPUOJKUXFUKN-UHFFFAOYSA-N
Synonym 3-bromomethyl pyridine hydrobromide, 3-picolyl bromide hydrobromide, 3-bromomethylpyridine hydrobromide, 3-bromomethyl-pyridine hydrobromide, pyridine, 3-bromomethyl-, hydrobromide, pubchem17664, acmc-209kf1, 3-brommethyl pyridine-hbr, 3-bromomethyl pyridine, bromide, 3-bromomethyl pyridinehydrobromide
PubChem CID 12707037
IUPAC Name 3-(bromomethyl)pyridine;hydrobromide
SMILES C1=CC(=CN=C1)CBr.Br

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 252.94
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Bromomethyl)pyridine Hydrobromide