accessibility menu, dialog, popup

UserName

CAS RN 4856-87-5

IUPAC Name: 1-[6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexyl]-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1,6-bismaleimidohexane
Molecular Weight (g/mol): 276.29
Molecular Formula: C14H16N2O4
InChi Key: PYVHLZLQVWXBDZ-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1CCCCCCN1C(=O)C=CC1=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)