missing translation for 'onlineSavingsMsg'
Learn More

1,6-Bismaleimidohexane, 97%

Catalog No. AAH58874ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58874ME 500 mg
AAH58874MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH58874ME Supplier Thermo Scientific Chemicals Supplier No. H58874ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4856-87-5 | C14H16N2O4 | 276.292 g/mol

A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.

Solubility
Soluble in chloroform.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 4856-87-5
Molecular Formula C14H16N2O4
Molecular Weight (g/mol) 276.292
MDL Number MFCD00047122
InChI Key PYVHLZLQVWXBDZ-UHFFFAOYSA-N
Synonym 1,6-bismaleimidohexane, 1,6-dimaleimidohexane, n,n'-hexamethylenedimaleimide, 1,6-bis maleimido hexane, 1,1'-hexane-1,6-diyl bis 1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1,1'-1,6-hexanediyl bis, maleimide, n,n'-hexamethylenedi, n,n'-hexamethylenebis maleimide, 1,1'-1,6-hexanediyl bis-1h-pyrrole-2,5-dione
PubChem CID 20992
IUPAC Name 1-[6-(2,5-dioxopyrrol-1-yl)hexyl]pyrrole-2,5-dione
SMILES C1=CC(=O)N(C1=O)CCCCCCN2C(=O)C=CC2=O

Specifications

Melting Point 144°C
Quantity 500 mg
Solubility Information Soluble in chloroform.
Formula Weight 276.29
Percent Purity 97%
Chemical Name or Material 1,6-Bismaleimidohexane
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-449-1
RTECSNumber ON5520000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only