missing translation for 'onlineSavingsMsg'
Learn More

1,6-Bis(maleimido)hexane 97.0+%, TCI America™

Catalog No. B1787100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1787100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1787100MG Supplier TCI America Supplier No. B1787100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4856-87-5
Molecular Formula C14H16N2O4
Molecular Weight (g/mol) 276.292
MDL Number MFCD00047122
InChI Key PYVHLZLQVWXBDZ-UHFFFAOYSA-N
Synonym 1,6-bismaleimidohexane, 1,6-dimaleimidohexane, n,n'-hexamethylenedimaleimide, 1,6-bis maleimido hexane, 1,1'-hexane-1,6-diyl bis 1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1,1'-1,6-hexanediyl bis, maleimide, n,n'-hexamethylenedi, n,n'-hexamethylenebis maleimide, 1,1'-1,6-hexanediyl bis-1h-pyrrole-2,5-dione
PubChem CID 20992
IUPAC Name 1-[6-(2,5-dioxopyrrol-1-yl)hexyl]pyrrole-2,5-dione
SMILES C1=CC(=O)N(C1=O)CCCCCCN2C(=O)C=CC2=O

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 100 mg
Formula Weight 276.29
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Bis(maleimido)hexane
RTECSNumber ON5520000
TSCA No