accessibility menu, dialog, popup

UserName

CAS RN 483367-10-8

NNNNNHNOO

CAS RN 483367-10-8

IUPAC Name: 9-cyclohexyl-N-[4-(morpholin-4-yl)phenyl]-2-(naphthalen-1-yloxy)-9H-purin-6-amine
Synonyms: purmorphamine
Molecular Weight (g/mol): 520.64
Molecular Formula: C31H32N6O2
InChi Key: FYBHCRQFSFYWPY-UHFFFAOYSA-N
SMILES: C1CCC(CC1)N1C=NC2=C(NC3=CC=C(C=C3)N3CCOCC3)N=C(OC3=C4C=CC=CC4=CC=C3)N=C12
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results