CAS RN 480-64-8
CAS RN 480-64-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
o-Orsellinic acid hydrate, 98%
CAS: 480-64-8 Molecular Formula: C8H8O4 Molecular Weight (g/mol): 168.15 MDL Number: MFCD16661187 InChI Key: AMKYESDOVDKZKV-UHFFFAOYSA-N Synonym: orsellinic acid,orsellic acid,o-orsellinic acid,orcinolcarboxylic acid,4,6-dihydroxy-o-toluic acid,unii-11xla0494b,benzoic acid, 2,4-dihydroxy-6-methyl,2,4-dihydroxy-6-methyl-benzoic acid,orsellinate,4,6-dihydroxy-2-methylbenzoic acid PubChem CID: 68072 ChEBI: CHEBI:32807 IUPAC Name: 2,4-dihydroxy-6-methylbenzoic acid SMILES: CC1=CC(O)=CC(O)=C1C(O)=O