missing translation for 'onlineSavingsMsg'
Learn More

o-Orsellinic acid hydrate, 98%, Thermo Scientific™

Catalog No. AC453290050
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453290050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC453290050 Supplier Thermo Scientific Chemicals Supplier No. 453290050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 480-64-8
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C8H8O4
MDL Number MFCD00210536,MFCD16661187
Quantity 5g
Synonym orsellinic acid, orsellic acid, o-orsellinic acid, orcinolcarboxylic acid, 4,6-dihydroxy-o-toluic acid, unii-11xla0494b, benzoic acid, 2,4-dihydroxy-6-methyl, 2,4-dihydroxy-6-methyl-benzoic acid, orsellinate, 4,6-dihydroxy-2-methylbenzoic acid
InChI Key AMKYESDOVDKZKV-UHFFFAOYSA-N
SMILES CC1=CC(O)=CC(O)=C1C(O)=O
IUPAC Name 2,4-dihydroxy-6-methylbenzoic acid
Molecular Weight (g/mol) 168.15
PubChem CID 68072
ChEBI CHEBI:32807
Formula Weight 186.17
Percent Purity 98%
Chemical Name or Material o-Orsellinic acid monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only