accessibility menu, dialog, popup

UserName

CH3OOOHOHO

CAS RN 480-44-4

IUPAC Name: 5,7-dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one
Synonyms: acacetin acacetine 4'-methoxyapigenin apigenin 4'-methyl ether apisenin 4'-methyl ether linarigenin buddleoflavonol akatsetin apigenin 4'-dimethyl ether linarisenin
Molecular Weight (g/mol): 284.27
Molecular Formula: C16H12O5
InChi Key: DANYIYRPLHHOCZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)